4-Chloro-2,6-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16216-25G 16216-5G
76925-686EA 289.63 USD
76925-686 76925-688
4-Chloro-2,6-difluorobenzaldehyde ≥97%
4-Chloro-2,6-difluorobenzaldehyde

Caution: for Research Use Only

Formula: C₇H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD03094460
CAS Number: 252004-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR