2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzonitrile4-Cyano-3,5-difluorobenzeneboronic acid pinacol ester

18331-500MG 18331-25G 18331-1G 18331-5G
76897-162EA 135.8 USD
76897-162 76897-164 76897-166 76897-168
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥95%
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Formula: C₁₃H₁₄BF₂NO₂
MW: 265.067 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22419275
CAS Number: 1003298-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR