2-Bromo-3-(trifluoromethyl)benzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
15185-1G
15185-25G
15185-5G
76895-756EA
62.6
USD
76895-756
76895-758
76895-760
2-Bromo-3-(trifluoromethyl)benzoic acid ≥95%
2-Bromo-3-(trifluoromethyl)benzoic acid
Caution: for Research Use Only
Formula:
C₈H₄BrF₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08532493 CAS Number: 177420-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...