2'-Fluoro-3'-methyl-3,4'-bipyridine ≥95% research grade

Supplier: AOBChem USA

124362-1G 124362-25G 124362-500MG 124362-5G
76894-854EA 153.84 USD
76894-854 76894-856 76894-858 76894-860
2'-Fluoro-3'-methyl-3,4'-bipyridine ≥95% research grade
2'-Fluoro-3'-methyl-3,4'-bipyridine
Formula: C₁₁H₉FN₂
CAS Number: 2918949-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR