4-(Difluoromethoxy)-3-fluorobenzaldehyde ≥95% research grade

Supplier: AOBChem USA

Warning

36497-1G 36497-25G 36497-500MG 36497-5G
76915-606EA 439.22 USD
76915-606 76915-608 76915-610 76915-612
4-(Difluoromethoxy)-3-fluorobenzaldehyde ≥95% research grade
4-(Difluoromethoxy)-3-fluorobenzaldehyde
Formula: C₈H₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11847153
CAS Number: 1214379-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR