3-Chloro-5-fluoro-4-methoxybenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

32292-1G 32292-25G 32292-5G
76914-168EA 62.6 USD
76914-168 76914-170 76914-172
3-Chloro-5-fluoro-4-methoxybenzyl alcohol ≥95%
3-Chloro-5-fluoro-4-methoxybenzyl alcohol

Caution: for Research Use Only

Formula: C₈H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD04115867
CAS Number: 886497-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR