3-Fluoro-2-methylbenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3,2-Dioxaborolane, 2-(3-fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-, 2-(3-Fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Fluoro-2-methylphenylboronic acid pinacol ester

32166-1G 32166-25G 32166-500MG 32166-5G
76914-114EA 231.28 USD
76914-114 76914-116 76914-118 76914-120
3-Fluoro-2-methylbenzeneboronic acid pinacol ester ≥97%
3-Fluoro-2-methylbenzeneboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₃H₁₈BFO₂
Storage Temperature: Ambient
MDL Number: MFCD18729892
CAS Number: 1417301-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR