3-Iodophenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
31514-25G
31514-5G
76914-000EA
375.56
USD
76914-000
76914-002
3-Iodophenylboronic acid pinacol ester ≥97%
3-Iodophenylboronic acid pinacol ester
Caution: for Research Use Only
Formula:
C₁₂H₁₆BIO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09953498 CAS Number: 408492-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...