4-Benzyloxy-2-(trifluoromethoxy)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

29197-1G 29197-25G 29197-500MG 29197-5G
76913-198EA 231.28 USD
76913-198 76913-200 76913-202 76913-204
4-Benzyloxy-2-(trifluoromethoxy)phenylboronic acid ≥97% research grade
4-Benzyloxy-2-(trifluoromethoxy)phenylboronic acid
Formula: C₁₄H₁₂BF₃O₄
Storage Temperature: Ambient
MDL Number: MFCD28806022
CAS Number: 1562343-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR