4-Bromo-1-ethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

17058-25G 17058-5G
76908-740EA 310.85 USD
76908-740 76908-742
4-Bromo-1-ethyl-1H-pyrazole ≥97%
4-Bromo-1-ethyl-1H-pyrazole

Caution: for Research Use Only

Formula: C₅H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD08235252
CAS Number: 71229-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR