2-(2-Iodo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
Supplier: AOBChem USA
|
Warning
|
40751-1G
40751-25G
40751-5G
40751-500MG
76889-816EA
84.88
USD
76889-816
76889-818
76889-820
77650-586
2-(2-Iodo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-Iodo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Caution: for Research Use Only
Formula:
C₁₃H₁₈BIO₃ Storage Temperature: Ambient |
MDL Number:
MFCD17215809 CAS Number: 1256781-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...