1-Bromo-2-ethyl-4-methoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

29166-1G 29166-25G 29166-500MG 29166-5G
76886-634EA 124.13 USD
76886-634 76886-636 76886-638 76886-640
1-Bromo-2-ethyl-4-methoxybenzene ≥97% research grade
1-Bromo-2-ethyl-4-methoxybenzene
Formula: C₉H₁₁BrO
Storage Temperature: Refrigerator
MDL Number: MFCD11043764
CAS Number: 34881-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR