1-Bromo-3-cyclopentylbenzene ≥97% research grade

Supplier: AOBChem USA

Warning

29001-1G 29001-25G 29001-500MG 29001-5G
76886-606EA 439.22 USD
76886-606 76886-608 76886-610 76886-612
1-Bromo-3-cyclopentylbenzene ≥97% research grade
1-Bromo-3-cyclopentylbenzene
Formula: C₁₁H₁₃Br
Storage Temperature: Refrigerator
MDL Number: MFCD20486549
CAS Number: 19920-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR