2,3-Difluoro-6-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

28490-25G 28490-5G
76886-434EA 375.56 USD
76886-434 76886-436
2,3-Difluoro-6-methoxybenzyl alcohol ≥97%
2,3-Difluoro-6-methoxybenzyl alcohol

Caution: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04116021
CAS Number: 773871-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR