Vertex® Filter Pipette Tips
Supplier: SCIENTIFIC SPECIALTIES, INC.
4117NSFS 4137NSFS 4237NAFS 4237NBFS 4237NSFS 4337NSFS 4347NSFS 4937NSFS
76808-776CS 233.84 USD
76808-776 76808-764 76808-772 76808-774 76808-778 76808-768 76808-766 76808-770
Vertex® Filter Pipette Tips
Pipette Tips
Clean liquid release for maximum sample recovery and increased accuracy. Vertex® filter tips feature NoStick® resin that provides researchers with exceptional liquid handling control. Low-retention properties facilitate a much cleaner release, dispensing all of the sampe and improving pipetting accuracy.
  • Universal pipettor compatibility
  • High-clarify virgin polypropylene (meets FDA 21 CFR 177.1520 and USP Class VI)
  • NoStick® resin technology facilitates clean release
  • Filters provide a hydrophobic HDPE barrier that won't contaminate or impede airflow
More Product Information