Mineral oil light, Muri-Lube®, sterile

Supplier: Fresenius Kabi

25410 25402
68099-152PK 1048.07 USD
68099-152 68099-150
Mineral oil light, Muri-Lube®, sterile
Mineral oil light
MDL Number: MFCD00131611
CAS Number: 8042-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR