Sodium chloride 0.9% USP for irrigation
Supplier: BAXTER IV SOLUTIONS
Synonyms:
Halite
, Sea salt
, Saline
2B7231
68000-504CS
2170.13
USD
68000-504
Sodium chloride 0.9% USP for irrigation
Sodium chloride
Solutions for irrigation (for Preparation of Slushed Solution)
- mEq/L: Na 154, Cl 154
- mOsmol/L (calc): 308
- NDC Number: 0338-0051-44
- pH: 5.5 (4.5–7.0)
Formula:
NaCl MW: 58.44 g/mol |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...