Sodium chloride 0.9% USP for irrigation

Supplier: BAXTER IV SOLUTIONS

Synonyms: HaliteSea saltNaClSaline

2B7126
68000-496CS 281.08 USD
68000-496
Sodium chloride 0.9% USP for irrigation
Sodium chloride

Solutions for irrigation.


  • mEq/L: Na 154, Cl 154
  • mOsmol/L (calc): 308
  • NDC Number: 0338-0047-46
  • pH: 5.5 (4.5–7.0)

Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR