Water for injection (WFI)

Supplier: Fresenius Kabi

Synonyms: Dihydrogen oxide

918510 918520
68099-178CS 11652.06 USD
68099-178 68099-176 68099-180 68099-186
Water for injection (WFI)
Water

Sterile injections comply with USP standards and are bar coded for maximum traceability.


  • Preservative-free

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR