Sodium chloride 3% USP for injection
Supplier: BAXTER IV SOLUTIONS
Synonyms:
Halite
, Sea salt
, NaCl
, Saline
2B1353Q
68000-350CS
724.42
USD
68000-350
Sodium chloride 3% USP for injection
Sodium chloride
Sterile solutions for injection.
- Intravenous use
- Subcutaneous use
- mEq/L: Na 513, Cl 513
- mOsmol/L (calc): 1027
- NDC Number: 0338-0054-03
- pH: 5.5 (5.0–7.5)
Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.
Formula:
NaCl MW: 58.44 g/mol |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...