Water USP, sterile

Supplier: B BRAUN MEDICAL

Synonyms: Dihydrogen oxide

PIC™
R5001-01
68200-062CS 126.89 USD
68200-062
Water USP, sterile
Water

  • Ideal for sterile overhead irrigation
  • Designed for easy non-drip pouring
  • Not manufactured with PVC or DEHP
  • Lightweight, durable and shatterproof
  • NDC number: 00264-2101-10


Ready for immediate use off the shelf, these high-quality solution come in semi-rigid Plastic Irrigation Containers (PIC™).


Weight: 21.5 lbs.

Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR