Water USP for injection (WFI), sterile
Supplier: B BRAUN MEDICAL
Synonyms:
Dihydrogen oxide
Sterile Water for Injection USP is a clear, colorless, odorless liquid. It is sterile, hypotonic, nonpyrogenic, and contains no bacteriostatic or antimicrobial agents. It is a diluent or solvent suitable for intravascular injection after first having been made approximately isotonic by the addition of suitable solute.
- NDC number: 0264-7850-20
Sterile Water for Injection USP is also used as a diluent or solvent for other parenteral drugs. As such, it contributes to hydration when provided in parenteral drug and fluid therapy, after the introduction of suitable additives and/or mixture with suitable solutes to approximate isotonicity.
Not manufactured with latex, PVC or DEHP.
Formula:
H₂O MW: 18.02 g/mol |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...