SSIbio® Vertex®LE Rainin LTS Compatible Pipette Tips
Supplier: SCIENTIFIC SPECIALTIES, INC.
41L5N00S 41L7N00S 41L7NS0S 41L7NSFS 42L5N00S 42L7N00S 42L7NS0S 42L7NSFS 43L5N00S 43L7N00S 43L7NS0S 43L7NSFS 49L7N00S 49L7NS0S 49L7NSFS
76881-972CS 233.99 USD
76881-972 76881-974 76881-976 76881-978 76881-980 76881-982 76881-984 76881-986 76881-988 76881-990 76881-992 76881-994 76881-996 76881-998 76882-000
SSIbio® Vertex®LE Rainin LTS Compatible Pipette Tips
Pipette Tips
VertexLE LTS tips are designed to precisely fit your Rainin® LiteTouch® Pipettes. Available in a variety of options from LTS filter tips to reloads, VertexLE LTS are an economical, high quality alternative for your laboratory. Experience pipetting excellence with Vertex®LE - the perfect drop-in replacement for your LTS® pipettors.
  • Compatible with Rainin® single- and multi-channel LiteTouch® pipettors
  • NoStick® ultra low-retention technology maximizes precision and accuracy with LTS pipettors
  • Available in racked, sterile, filtered sterile and reload
More Product Information