miniPCR® miniX Thermal Cyclers
Supplier: AMPLYUS LLC
QP-1000-08 QP-1016-16 QP-1000-48 QP-1016-46
470352-024EA 815.19 USD
470352-024 470351-984 470355-082 470355-080
miniPCR® miniX Thermal Cyclers
Thermal Cyclers
These 8 or 16-well thermal cyclers offer a personalized PCR experience for maximum engagement at a budget-conscious price point. Small and portable, this fully-featured PCR machine fits on your bench and travels wherever your science goes.
  • Peer-reviewed published results
  • Durable in the lab, classroom, field, even outer space
  • Weighs under one pound, fits in the palm of your hand
  • AC or battery powered (battery not included)
  • Universal compatibility via USB or Bluetooth®
  • 3-year warranty and 30-day satisfaction guarantee
  • Fully compatible with standard PCR tubes
More Product Information