Anti-IgG4 Monoclonal Antibody [clone: R1] (Biotin)

Supplier: Sino Biological
HG4K-B-100 HG4K-B-25 HG4K-B-500
76777-226EA 4545.75 USD
76777-226 76777-200 76777-644
Anti-IgG4 Monoclonal Antibody [clone: R1] (Biotin)
Antibodies
The antibody was produced using recombinant antibody technology and then biotinylated.

Type: Control
Antigen: IgG4
Clonality: Monoclonal
Clone: R1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR