miniPCR® DNA Discovery System™
Supplier: AMPLYUS LLC
QP-2000-08 QP-2016-16
470352-080EA 1238.89 USD
470352-080 470352-070
miniPCR® DNA Discovery System™
Educational Classroom Kits and Activities
The complete biotech toolkit for your lab or classroom.
The miniPCR® thermal cycler features a heated lid, 8×0.2 ml PCR tube capacity (strip-compatible) with an option for 16 wells (upgrade to mini16X), and is fully compatible with standard tubes and reagents. Offers full control via free Windows, Mac, iOS, Chrome OS, or Android app.
The blueGel™ electrophoresis and visualization system uses 10x less reagents than conventional electrophoresis and employs safe blue-light transilluminator technology with no need for ethidium bromide, UV, or high voltage.
Certifications: miniPCR® and blueGel™ are made in the USA and CE marked.
Delivery information: miniPCR DNA Discovery System™ includes: miniPCR® mini8X or mini16X thermal cycler; blueGel™ electrophoresis system; 2 to 20 μl micropipette; 2-year Warranty.