MAXgene® GMP Transfection Reagents
Supplier: KYFORA BIO LLC.
GMP grade transfection reagent for eukaryotic cells.
- High transfection efficiency
- Low cytotoxicity
- Reliable performance
- Flexible workflow
- Compatible for viral vector and protein production
- Fully synthetic
- Adherent and suspension cells
- DNA and protein delivery
MAXgene® GMP Transfection Reagent is a cGMP transfection reagent for the development and manufacturing of viral vectors for cell-and gene-based therapies. It is an ideal reagent in HEK293 and CHO systems for the manufacture of AAVs, LVs and recombinant proteins. MAXgene® GMP capitalizes on the efficiency and scalability of Polysciences’ PEI MAX® while adding the validation process and regulatory components necessary for moving into clinical and commercial manufacturing. It is manufactured in accordance with cGMP under an ISO 13485 Quality Management System. Please contact us at info@polysciences.com for MAXgene produced in accordance with 21 CFR 210, 211.