HyClone™ SFM4MAb media, Cytiva
Supplier: Cytiva
SH30513.02 SH30535.02
14224-300EA 97.63 USD
14224-300 76462-528
HyClone™ SFM4MAb media, Cytiva
Cell Culture Media
HyClone™ SFM4MAb media was developed through the Metabolic Pathway Design approach to increase process yields for manufacturing of human and humanised recombinant antibodies for therapeutic use.