Guardian 3000 Digital Hot Plate Stirrers
Supplier: OHAUS CORP.
30680266 30680267 30680268
470351-004EA 613.6 USD
470351-004 470351-002 470351-000
Guardian 3000 Digital Hot Plate Stirrers
Stirrers Magnetic Hotplate Stirrers
Advanced, digital hotplate stirrers for all your laboratory heating and stirring needs.
  • SafetyHeat™ early overtemperature detection system
  • Rugged, spill-resistant metal housing
  • Chemical-resistant ceramic top
  • 2 years warranty
These hotplate stirrers are designed for economical heating and stirring in all laboratory settings. The bright LCD display includes easy-to-read temperature and speed settings, intuitive icons and a prominent hot top indicator that lights up when the heater is above 40 °C. Select from three plate sizes to suit your application. All models can be used with the optional temperature probe for precise temperature control of the sample.
Certifications: TUV.