2,4,7,8,9-Penta-O-acetyl-N-acetylneuraminic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2R)-1-((2R,3R,4S)-3-Acetamido-4,6-diacetoxy-6-(methoxycarbonyl)tetrahydro-2H-pyran-2-yl)propane-1,2,3-triyl triacetate, N-Acetylneuraminic acid methyl ester 2,4,7,8,9-Pentaacetate, Methyl 5-Acetamido-2,4,7,8,9-penta-O-acetyl-3,5-dideoxy-D-glycero-D-galacto-2-nonulopyranosonate

A869059-5G A869059-1G
76880-664EA 182 USD
76880-664 77886-867
2,4,7,8,9-Penta-O-acetyl-N-acetylneuraminic acid methyl ester 98%
2,4,7,8,9-Penta-O-acetyl-N-acetylneuraminic acid methyl ester

Caution: for Research Use Only

Formula: C₂₂H₃₁NO₁₄
MW: 533.48 g/mol
MDL Number: MFCD30342209
CAS Number: 73208-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR