FR180204 98%

Supplier: Ambeed
Warning

Synonyms: FR180204FR 180204FR-1802045-(2-phenylpyrazolo[1,5-a]pyridin-3-yl)-2H-pyrazolo[3,4-c]pyridazin-3-amine5-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-c]pyridazin-3-amine

A867944-1MG A867944-10MG A867944-5MG
76880-642EA 177.18 USD
76880-642 76880-644 76880-646
FR180204 98%
FR180204

Caution: for Research Use Only

Formula: C₁₈H₁₃N₇
MW: 327.34 g/mol
MDL Number: MFCD14585805
CAS Number: 865362-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR