5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-4,5-(difluoromethylenedioxy)aniline, 6-Bromo-2,2-difluorobenzo[d][1,3]dioxol-5-amine, 6-Bromo-2,2-difluoro-1,3-benzodioxol-5-amine
A444194-100MG
A444194-1G
A444194-250MG
A444194-5G
76877-416EA
9
USD
76877-416
76877-418
76877-420
76877-422
5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole 97%
5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole
Caution: for Research Use Only
Formula:
C₇H₄BrF₂NO₂ |
MDL Number:
MFCD04972651 CAS Number: 887267-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...