2-[(Acetylthio)methyl]-3-phenylpropionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-[(Acetylthio)methyl]-3-phenylpropionic Acid

A427227-100G A427227-10G A427227-25G
76877-224EA 104 USD
76877-224 76877-226 76877-228 76877-230
2-[(Acetylthio)methyl]-3-phenylpropionic acid 97%
2-[(Acetylthio)methyl]-3-phenylpropionic acid

Caution: for Research Use Only

Formula: C₁₂H₁₄O₃S
MW: 238.30 g/mol
MDL Number: MFCD00236751
CAS Number: 91702-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR