2-Amino-5-(trifluoromethoxy)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-amino-5-(trifluoromethoxy)benzoic acid
A377097-1G
A377097-250MG
A377097-25G
A377097-5G
76876-606EA
11
USD
76876-606
76876-608
76876-610
76876-612
2-Amino-5-(trifluoromethoxy)benzoic acid 97%
2-Amino-5-(trifluoromethoxy)benzoic acid
Caution: for Research Use Only
Formula:
C₈H₆F₃NO₃ MW: 221.14 g/mol |
MDL Number:
MFCD00176651 CAS Number: 83265-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...