2-Amino-3-bromobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A196798-100MG
A196798-10G
A196798-1G
A196798-250MG
A196798-5G
A196798-25G
76873-254EA
14
USD
76873-254
76873-256
76873-258
76873-260
76873-262
77883-423
2-Amino-3-bromobenzaldehyde 97%
2-Amino-3-bromobenzaldehyde
Caution: for Research Use Only
Formula:
C₇H₆BrNO MW: 200.033 g/mol |
MDL Number:
MFCD18389339 CAS Number: 145123-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...