4-Fluoro-2-methylpyridine 97%

Supplier: Ambeed
Warning

A183081-1G A183081-5G A183081-250MG
76872-812EA 63 USD
76872-812 76872-814 77883-380
4-Fluoro-2-methylpyridine 97%
4-Fluoro-2-methylpyridine

Caution: for Research Use Only

Formula: C₆H₆FN
MDL Number: MFCD04114145
CAS Number: 766-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR