6-Amino-1-naphthalenesulfonic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Amino-1-naphthalenesulphonic acid, 6-Amino-1-naphthalenesulfonic acid
A152315-100G
A152315-25G
A152315-5G
76871-644EA
83
USD
76871-644
76871-646
76871-648
6-Amino-1-naphthalenesulfonic acid 95%
6-Amino-1-naphthalenesulphonic acid
Caution: for Research Use Only
Formula:
C₁₀H₉NO₃S MW: 223.2500 g/mol |
MDL Number:
MFCD00035724 CAS Number: 81-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...