2-Amino-5-bromo-3-methylbenzoic acid 97%

Supplier: Ambeed
Warning

A145539-10G A145539-1G A145539-250MG A145539-25G A145539-5G
76871-428EA 95 USD
76871-428 76871-430 76871-432 76871-434 76871-436
2-Amino-5-bromo-3-methylbenzoic acid 97%
2-Amino-5-bromo-3-methylbenzoic acid

Caution: for Research Use Only

Formula: C₈H₈BrNO₂
MDL Number: MFCD05664860
CAS Number: 206548-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR