2,4-Difluoro-5-nitrophenol 97%
Supplier: Ambeed
|
Danger
|
A138028-1G
A138028-10G
A138028-5G
A138028-100G
A138028-250MG
A138028-25G
76871-040EA
152.77
USD
76871-040
76871-034
76871-042
76871-032
76871-038
76871-036
2,4-Difluoro-5-nitrophenol 97%
2,4-Difluoro-5-nitrophenol
Caution: for Research Use Only
Formula:
C₆H₃F₂NO₃ MW: 175.09 g/mol |
MDL Number:
MFCD08443942 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...