4-Methylumbelliferone 99%

Supplier: Ambeed
Warning

A132122-1000G A132122-100G A132122-10G A132122-25G A132122-25MG A132122-500G
76870-784EA 148 USD
76870-784 76870-786 76870-788 76870-790 76870-792 76870-794
4-Methylumbelliferone 99%
4-Methylumbelliferone

Caution: for Research Use Only

Formula: C₁₀H₈O₃
MDL Number: MFCD00006866
CAS Number: 90-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR