Pyrrolidin-3-ol hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (±)-3-Hydroxypyrrolidine hydrochloride

A131822-1G A131822-5G A131822-25G A131822-100G
76870-780EA 5 USD
76870-780 76870-782 77670-864 77883-061
Pyrrolidin-3-ol hydrochloride 98%
Pyrrolidin-3-ol hydrochloride

Caution: for Research Use Only

Formula: C4H10ClNO
MDL Number: MFCD03001754
CAS Number: 86070-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR