6-Aminocaproic acid 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
ε-Amino-n-caproic acid
, H-Eahx-OH
, Aminocaproic Acid
, 6-Aminocaproic Acid
, 6-Aminohexanoic acid
A449367-100G
A449367-10G
A449367-500G
A449367-25G
77034-080EA
73.13
USD
77034-080
77034-076
77034-078
76842-662
6-Aminocaproic acid 98+%
6-Aminocaproic acid
Caution: for Research Use Only
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol |
MDL Number:
MFCD00008238 CAS Number: 60-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...