L(-)-Tryptophan 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-α-Amino-3-indolepropionic acid
, (S)-2-Amino-3-(3-indolyl)propionic acid
, H-Trp-OH
, L-Tryptophan
, Tryptophan
A317029-500G
A317029-25G
A317029-100G
A317029-10G
76838-908EA
8.24
USD
76838-908
77114-676
77114-678
77114-674
L(-)-Tryptophan 98%
L(-)-Tryptophan
Caution: for Research Use Only
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...