Ethyl-3-acetylbenzoate 98%
Supplier: Ambeed
|
Warning
|
A797401-1G
A797401-250MG
A797401-5G
A797401-25G
76832-292EA
22
USD
76832-292
76832-294
76832-296
77886-776
Ethyl-3-acetylbenzoate 98%
Ethyl-3-acetylbenzoate
Caution: for Research Use Only
Formula:
C₁₁H₁₂O₃ |
MDL Number:
MFCD02261283 CAS Number: 37847-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...