2-(2-Chlorophenyl)-2-hydroxyacetonitrile 95%

Supplier: Ambeed
Warning

A713653-100MG A713653-1G A713653-250MG
76831-720EA 74 USD
76831-720 76831-722 76831-724
2-(2-Chlorophenyl)-2-hydroxyacetonitrile 95%
2-(2-Chlorophenyl)-2-hydroxyacetonitrile

Caution: for Research Use Only

Formula: C₈H₆ClNO
MDL Number: MFCD12169318
CAS Number: 13312-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR