3-Bromo-4-propylbenzoic acid 95+%

Supplier: Ambeed
Warning

A692408-100MG A692408-250MG
76831-570EA 48 USD
76831-570 76831-572
3-Bromo-4-propylbenzoic acid 95+%
3-Bromo-4-propylbenzoic acid

Caution: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD11110861
CAS Number: 1131615-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR