7-Bromo-2-methyl-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Ambeed
Warning

A581941-100MG A581941-1G A581941-250MG A581941-50MG A581941-5G
76830-782EA 38 USD
76830-782 76830-784 76830-786 76830-788 77886-414
7-Bromo-2-methyl-1,2,3,4-tetrahydroisoquinoline 95%
7-Bromo-2-methyl-1,2,3,4-tetrahydroisoquinoline

Caution: for Research Use Only

Formula: C₁₀H₁₂BrN
MDL Number: MFCD15474923
CAS Number: 258515-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR