Glyoxylic acid monohydrate 95%

Supplier: Ambeed
Danger

Synonyms: Formylformic acid monohydrateOxoacetic acid monohydrate2-Oxoethanoic acid monohydrate2,2-Dihydroxyacetic acid monohydrate

A432964-100G A432964-25G
77613-340EA 82.4 USD
77613-340 76829-562
Glyoxylic acid monohydrate 95%
Glyoxylic acid monohydrate

Caution: for Research Use Only

Formula: C₂H₂O₃·1H₂O
MW: 92.05 g/mol
MDL Number: MFCD00127974
CAS Number: 563-96-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR