TPCA-1 98+%

Supplier: Ambeed
Warning

Synonyms: 5-(4-Fluorophenyl)-2-ureidothiophene-3-carboxamide, 2-[(aminocarbonyl)amino]-5-(4-fluorophenyl)-3-thiophenecarboxamide, IKK2 Inhibitor IV

A377370-10MG A377370-1MG A377370-50MG A377370-5MG
76828-962EA 49 USD
76828-962 76828-964 76828-966 76828-968
TPCA-1 98+%
TPCA-1

Caution: for Research Use Only

Formula: C₁₂H₁₀FN₃O₂S
MW: 279.3 g/mol
MDL Number: MFCD09037541
CAS Number: 507475-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR