7-Acetyl-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(1H-Indazol-7-yl)ethan-1-one1-(1H-Indazol-7-yl)ethanone

A225670-1G A225670-5G A225670-250MG
76826-286EA 39.26 USD
76826-286 76826-288 76826-284
7-Acetyl-1H-indazole 98%
7-Acetyl-1H-indazole

Caution: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.173 g/mol
MDL Number: MFCD11869765
CAS Number: 1159511-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR